Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IPO11
Entrez Gene ID
538236
Gene Name
importin 11
Gene Aliases
-
Chromosome Location
Chr.20: 16791868 - 16995959 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192932.2 NP_001179861.2 3 - 4 311 91

View Full Product Details