Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC35F3
Entrez Gene ID
538365
Gene Name
solute carrier family 35 member F3
Gene Aliases
-
Chromosome Location
Chr.28: 6720734 - 7155385 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192935.2 NP_001179864.2 1 - 2 51 86

View Full Product Details