Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYSMD3
Entrez Gene ID
613760
Gene Name
LysM domain containing 3
Gene Aliases
-
Chromosome Location
Chr.7: 90083735 - 90097651 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192982.1 NP_001179911.1 1 - 2 258 88

View Full Product Details