Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CMBL
Entrez Gene ID
516405
Gene Name
carboxymethylenebutenolidase homolog
Gene Aliases
-
Chromosome Location
Chr.20: 62947984 - 62973176 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192983.3 NP_001179912.1 1 - 2 159 64
GenBank mRNA BC102137.1 - 1 - 2 133 64

View Full Product Details