Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CIAO2B
Entrez Gene ID
614585
Gene Name
cytosolic iron-sulfur assembly component 2B
Gene Aliases
FAM96B
Chromosome Location
Chr.18: 34635125 - 34638372 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193038.1 NP_001179967.1 3 - 4 348 75

View Full Product Details