Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCYT1B
Entrez Gene ID
526175
Gene Name
phosphate cytidylyltransferase 1, choline, beta
Gene Aliases
-
Chromosome Location
Chr.X: 118649793 - 118796606 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193051.1 NP_001179980.1 2 - 3 222 87

View Full Product Details