Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLA2G5
Entrez Gene ID
614911
Gene Name
phospholipase A2 group V
Gene Aliases
-
Chromosome Location
Chr.2: 132644532 - 132665181 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193052.1 NP_001179981.1 5 - 6 605 80

View Full Product Details