Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GSAP
Entrez Gene ID
615147
Gene Name
gamma-secretase activating protein
Gene Aliases
PION
Chromosome Location
Chr.4: 43815335 - 43913877 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193061.1 NP_001179990.1 22 - 23 1763 124

View Full Product Details