Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC4D
Entrez Gene ID
616646
Gene Name
C-type lectin domain family 4 member D
Gene Aliases
-
Chromosome Location
Chr.5: 101685217 - 101693399 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193117.1 NP_001180046.1 3 - 4 418 70
GenBank mRNA FE015823.1 - 3 - 4 418 70

View Full Product Details