Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MPC2
Entrez Gene ID
616718
Gene Name
mitochondrial pyruvate carrier 2
Gene Aliases
BRP44
Chromosome Location
Chr.3: 752398 - 783103 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193121.1 NP_001180050.1 4 - 5 334 118
GenBank mRNA BC109938.1 - 3 - 4 547 118

View Full Product Details