Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RUNX3
Entrez Gene ID
617389
Gene Name
RUNX family transcription factor 3
Gene Aliases
-
Chromosome Location
Chr.2: 127998389 - 128062834 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193158.2 NP_001180087.2 4 - 5 890 100
NM_001369867.1 NP_001356796.1 6 - 7 1017 100

View Full Product Details