Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH2D1B
Entrez Gene ID
617485
Gene Name
SH2 domain containing 1B
Gene Aliases
-
Chromosome Location
Chr.3: 7067290 - 7097285 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193162.1 NP_001180091.1 3 - 4 492 81

View Full Product Details