Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OTX2
Entrez Gene ID
538953
Gene Name
orthodenticle homeobox 2
Gene Aliases
ORX2
Chromosome Location
Chr.10: 69215035 - 69224752 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193201.1 NP_001180130.1 1 - 2 98 66

View Full Product Details