Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AP3M2
Entrez Gene ID
509427
Gene Name
adaptor related protein complex 3 subunit mu 2
Gene Aliases
-
Chromosome Location
Chr.27: 37031648 - 37050493 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193240.2 NP_001180169.1 6 - 7 903 80

View Full Product Details