Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARCN1
Entrez Gene ID
533078
Gene Name
archain 1
Gene Aliases
-
Chromosome Location
Chr.15: 29227702 - 29256278 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195013.1 NP_001181942.1 1 - 2 151 71

View Full Product Details