Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
TEX43
Entrez Gene ID
100126817
Gene Name
testis expressed 43
Gene Aliases
C7H5orf48
Chromosome Location
Chr.7: 27259101 - 27264651 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195070.1 NP_001181999.1 2 - 3 190 73

View Full Product Details