Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FASTKD3
Entrez Gene ID
513260
Gene Name
FAST kinase domains 3
Gene Aliases
-
Chromosome Location
Chr.20: 65343151 - 65355022 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024528.2 NP_001019699.2 2 - 3 1565 104

View Full Product Details