Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRIP1
Entrez Gene ID
574093
Gene Name
cysteine rich protein 1
Gene Aliases
-
Chromosome Location
Chr.21: 69755669 - 69757183 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025349.2 NP_001020520.1 - 99 69
GenBank mRNA AY911323.1 - 1 - 2 101 69
BC102754.1 - - 99 69

View Full Product Details