Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDOR1
Entrez Gene ID
513873
Gene Name
NADPH dependent diflavin oxidoreductase 1
Gene Aliases
-
Chromosome Location
Chr.11: 106224779 - 106235646 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046158.2 NP_001039623.2 3 - 4 400 85
GenBank mRNA BC112851.1 - 3 - 4 400 85
BT025363.1 - 3 - 4 328 85
DV838559.1 - 3 - 4 400 85

View Full Product Details