Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRM7
Entrez Gene ID
535362
Gene Name
glutamate metabotropic receptor 7
Gene Aliases
-
Chromosome Location
Chr.22: 18627109 - 19567631 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076055.3 NP_001069523.3 8 - 9 2669 84
GenBank mRNA AW315455.1 - 1 - 2 278 84
DV871711.1 - 1 - 2 314 84

View Full Product Details