Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EVA1C
Entrez Gene ID
516536
Gene Name
eva-1 homolog C
Gene Aliases
C1H21orf63
Chromosome Location
Chr.1: 2959342 - 3081902 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199027.1 NP_001185956.1 7 - 8 953 72
GenBank mRNA BC126735.1 - 6 - 7 860 72

View Full Product Details