Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KATNBL1
Entrez Gene ID
540295
Gene Name
katanin regulatory subunit B1 like 1
Gene Aliases
C10H15orf29
Chromosome Location
Chr.10: 28406090 - 28451549 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199064.3 NP_001185993.1 7 - 8 852 95

View Full Product Details