Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MLXIPL
Entrez Gene ID
788534
Gene Name
MLX interacting protein like
Gene Aliases
-
Chromosome Location
Chr.25: 33576946 - 33632423 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205408.1 NP_001192337.1 2 - 3 416 66

View Full Product Details