Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPR88
Entrez Gene ID
781302
Gene Name
G protein-coupled receptor 88
Gene Aliases
-
Chromosome Location
Chr.3: 42742344 - 42744302 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205412.1 NP_001192341.1 1 - 2 379 76

View Full Product Details