Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC281376
Entrez Gene ID
281376
Gene Name
serpin family B member 2-like
Gene Aliases
PAI2
Chromosome Location
Chr.24: 12446966 - 12464311 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192079.1 NP_001179008.1 3 - 4 340 108

View Full Product Details