Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSPB3
Entrez Gene ID
616007
Gene Name
heat shock protein family B (small) member 3
Gene Aliases
-
Chromosome Location
Chr.20: 24638174 - 24638869 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046570.2 NP_001040035.1 1 - 1 613 107
GenBank mRNA BC112874.1 - 1 - 1 614 107

View Full Product Details