Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRRN2
Entrez Gene ID
767860
Gene Name
leucine rich repeat neuronal 2
Gene Aliases
LRRN5
Chromosome Location
Chr.16: 2358404 - 2361430 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076971.1 NP_001070439.1 2 - 2 1123 126
GenBank mRNA BC142306.1 - 2 - 2 2840 126
BT021610.1 - 2 - 2 1123 126

View Full Product Details