Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SNHG12
- Entrez Gene ID
-
100174923
- Gene Name
- small nucleolar RNA host gene 12 (non-protein coding)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 124850784 - 124854008 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_038147.1 |
|
2 - 3 |
756 |
79 |
GenBank mRNA |
EU848421.1 |
- |
2 - 3 |
757 |
79 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
TATA-box binding protein associated factor 12 |
TAF12 |
|
-17.6kb
|
Sense |
Chr. - 124810106 - 124834314 on Build ARS-UCD1.2 |
Downstream |
tRNA selenocysteine 1 associated protein 1 |
TRNAU1AP |
TRSPAP1 |
1.5kb
|
Anti-sense |
Chr. - 124854126 - 124878715 on Build ARS-UCD1.2 |
View Full Product Details