Product Details
- SNP ID
-
rs76353298
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:241547429 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAGCAGGCAGAGGCCAGCAAAGCA[C/T]GCACTGGAGTCCACAGGCGCTGATC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BOK-AS1
PubMed Links
Gene Details
- Gene
- BOK-AS1
- Gene Name
- BOK antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details