Product Details

SNP ID
rs76688672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:194137822 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACTCGGCTGCTCGGCCACCTGGC[C/T]AACTGCATGACCCAGATCAATGCCA
Phenotype
MIM: 139605
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HES1 PubMed Links

Gene Details

Gene
HES1
Gene Name
hes family bHLH transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005524.3 671 Silent Mutation GCC,GCT A144A NP_005515.1

View Full Product Details