Product Details
- SNP ID
-
rs77444380
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:111047127 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAACTACTGGCCTATCACTGAAGA[A/G]TATCTAAAGGACTATCAATCAAGTC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PVRL3-AS1
PubMed Links
Gene Details
- Gene
- PVRL3-AS1
- Gene Name
- PVRL3 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details