Product Details
- SNP ID
-
rs79024054
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:112536061 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCACTAATCAATAAAAGAAAGCAAA[C/T]AGGTCACCTAGAAAAATCACAAGGT
- Phenotype
-
MIM: 609606
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ATG3
PubMed Links
Gene Details
- Gene
- ATG3
- Gene Name
- autophagy related 3
View Full Product Details