Product Details

SNP ID
rs75476744
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:118901541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCAGGTGCTGCCCTCATTAATAG[C/T]AACCTACTGTAATAGCAAGGGAGAT
Phenotype
MIM: 608351
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGSF11 PubMed Links

Gene Details

Gene
IGSF11
Gene Name
immunoglobulin superfamily member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015887.1 2522 UTR 3 NP_001015887.1
NM_152538.2 2522 UTR 3 NP_689751.2
XM_006713517.1 2522 UTR 3 XP_006713580.1
XM_011512462.2 2522 UTR 3 XP_011510764.1
XM_011512463.1 2522 UTR 3 XP_011510765.1
XM_011512464.2 2522 UTR 3 XP_011510766.1
XM_011512465.1 2522 UTR 3 XP_011510767.1
XM_011512466.2 2522 UTR 3 XP_011510768.1
XM_011512467.2 2522 UTR 3 XP_011510769.1
XM_011512468.1 2522 UTR 3 XP_011510770.1
XM_011512469.1 2522 UTR 3 XP_011510771.1
XM_011512470.1 2522 UTR 3 XP_011510772.1
XM_011512471.2 2522 UTR 3 XP_011510773.1
XM_011512472.1 2522 UTR 3 XP_011510774.1
XM_011512474.2 2522 Intron XP_011510776.1
XM_017005789.1 2522 UTR 3 XP_016861278.1
XM_017005790.1 2522 UTR 3 XP_016861279.1

View Full Product Details