Product Details

SNP ID
rs77476009
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:118901989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTACTGAATTTTGCTCATGTAAAA[A/G]GTTTTGATTATATGATAGAAGAGTC
Phenotype
MIM: 608351
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
IGSF11 PubMed Links

Gene Details

Gene
IGSF11
Gene Name
immunoglobulin superfamily member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015887.1 2074 UTR 3 NP_001015887.1
NM_152538.2 2074 UTR 3 NP_689751.2
XM_006713517.1 2074 UTR 3 XP_006713580.1
XM_011512462.2 2074 UTR 3 XP_011510764.1
XM_011512463.1 2074 UTR 3 XP_011510765.1
XM_011512464.2 2074 UTR 3 XP_011510766.1
XM_011512465.1 2074 UTR 3 XP_011510767.1
XM_011512466.2 2074 UTR 3 XP_011510768.1
XM_011512467.2 2074 UTR 3 XP_011510769.1
XM_011512468.1 2074 UTR 3 XP_011510770.1
XM_011512469.1 2074 UTR 3 XP_011510771.1
XM_011512470.1 2074 UTR 3 XP_011510772.1
XM_011512471.2 2074 UTR 3 XP_011510773.1
XM_011512472.1 2074 UTR 3 XP_011510774.1
XM_011512474.2 2074 Intron XP_011510776.1
XM_017005789.1 2074 UTR 3 XP_016861278.1
XM_017005790.1 2074 UTR 3 XP_016861279.1

View Full Product Details