Product Details

SNP ID
rs79780420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:121663319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGATCAGTCAGCCCACTTGTCTTC[C/T]CTCTTCTTTAGGGAGAGGCTAGGCA
Phenotype
MIM: 602500 MIM: 601306
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GOLGB1 PubMed Links

Gene Details

Gene
GOLGB1
Gene Name
golgin B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256486.1 11446 UTR 3 NP_001243415.1
NM_001256487.1 11446 UTR 3 NP_001243416.1
NM_001256488.1 11446 UTR 3 NP_001243417.1
NM_004487.4 11446 UTR 3 NP_004478.3
XM_005247371.4 11446 UTR 3 XP_005247428.1
XM_005247372.4 11446 UTR 3 XP_005247429.1
XM_005247373.1 11446 UTR 3 XP_005247430.1
XM_006713587.1 11446 UTR 3 XP_006713650.1
XM_006713588.1 11446 UTR 3 XP_006713651.1
XM_006713589.1 11446 UTR 3 XP_006713652.1
XM_006713590.1 11446 UTR 3 XP_006713653.1
XM_006713591.1 11446 UTR 3 XP_006713654.1
XM_011512699.2 11446 UTR 3 XP_011511001.1
XM_017006189.1 11446 UTR 3 XP_016861678.1
XM_017006190.1 11446 UTR 3 XP_016861679.1
XM_017006191.1 11446 UTR 3 XP_016861680.1
XM_017006192.1 11446 UTR 3 XP_016861681.1
XM_017006193.1 11446 UTR 3 XP_016861682.1
XM_017006194.1 11446 UTR 3 XP_016861683.1
XM_017006195.1 11446 UTR 3 XP_016861684.1
Gene
HCLS1
Gene Name
hematopoietic cell-specific Lyn substrate 1
There are no transcripts associated with this gene.

View Full Product Details