Product Details
- SNP ID
-
rs77221874
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:121669210 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGTCTCTCACCTTCTCTTTACTC[A/C]CCGCCTGCAGCTCTTGAACCTGCTT
- Phenotype
-
MIM: 602500
MIM: 601306
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GOLGB1
PubMed Links
Gene Details
- Gene
- GOLGB1
- Gene Name
- golgin B1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001256486.1 |
9798 |
Missense Mutation |
GGG,GTG |
G3108V |
NP_001243415.1 |
NM_001256487.1 |
9798 |
Intron |
|
|
NP_001243416.1 |
NM_001256488.1 |
9798 |
Intron |
|
|
NP_001243417.1 |
NM_004487.4 |
9798 |
Intron |
|
|
NP_004478.3 |
XM_005247371.4 |
9798 |
Missense Mutation |
GGG,GTG |
G3108V |
XP_005247428.1 |
XM_005247372.4 |
9798 |
Missense Mutation |
GGG,GTG |
G3069V |
XP_005247429.1 |
XM_005247373.1 |
9798 |
Missense Mutation |
GGG,GTG |
G3067V |
XP_005247430.1 |
XM_006713587.1 |
9798 |
Intron |
|
|
XP_006713650.1 |
XM_006713588.1 |
9798 |
Missense Mutation |
GGG,GTG |
G3103V |
XP_006713651.1 |
XM_006713589.1 |
9798 |
Missense Mutation |
GGG,GTG |
G3102V |
XP_006713652.1 |
XM_006713590.1 |
9798 |
Intron |
|
|
XP_006713653.1 |
XM_006713591.1 |
9798 |
Missense Mutation |
GGG,GTG |
G3028V |
XP_006713654.1 |
XM_011512699.2 |
9798 |
Missense Mutation |
GGG,GTG |
G3108V |
XP_011511001.1 |
XM_017006189.1 |
9798 |
Missense Mutation |
GGG,GTG |
G3103V |
XP_016861678.1 |
XM_017006190.1 |
9798 |
Missense Mutation |
GGG,GTG |
G3102V |
XP_016861679.1 |
XM_017006191.1 |
9798 |
Missense Mutation |
GGG,GTG |
G3067V |
XP_016861680.1 |
XM_017006192.1 |
9798 |
Missense Mutation |
GGG,GTG |
G3064V |
XP_016861681.1 |
XM_017006193.1 |
9798 |
Missense Mutation |
GGG,GTG |
G3063V |
XP_016861682.1 |
XM_017006194.1 |
9798 |
Missense Mutation |
GGG,GTG |
G3028V |
XP_016861683.1 |
XM_017006195.1 |
9798 |
Missense Mutation |
GGG,GTG |
G2855V |
XP_016861684.1 |
- Gene
- HCLS1
- Gene Name
- hematopoietic cell-specific Lyn substrate 1
There are no transcripts associated with this gene.
View Full Product Details