Product Details
- SNP ID
-
rs77580824
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:122325322 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACCTGGAGGCTTATCTGAGGCCAA[A/C]CCCGCCACTCCAGAAATCCAGGAGA
- Phenotype
-
MIM: 184600
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CSTA
PubMed Links
Gene Details
- Gene
- CSTA
- Gene Name
- cystatin A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005213.3 |
159 |
Missense Mutation |
AAA,AAC |
K10N |
NP_005204.1 |
View Full Product Details