Product Details

SNP ID
rs76659724
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:94072004 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCAGGTGTGTGCAAGGGGAACT[A/G]TAAGAATCATATTAATACACAGCTT
Phenotype
MIM: 616588
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHFR2 PubMed Links

Gene Details

Gene
DHFR2
Gene Name
dihydrofolate reductase 2
There are no transcripts associated with this gene.

Gene
NSUN3
Gene Name
NOP2/Sun RNA methyltransferase family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022072.3 Intron NP_071355.1

View Full Product Details