Product Details

SNP ID
rs78453588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:133801258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACTACAACTGGTTTCATTGTAAC[C/T]TTCCATCTGCTATTGGAATCAAGAA
Phenotype
MIM: 616883
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SRPRB PubMed Links

Gene Details

Gene
SRPRB
Gene Name
SRP receptor beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021203.3 Intron NP_067026.3

View Full Product Details