Product Details

SNP ID
rs78244234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:97838657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACCAAGAACGGTGCCTGACACA[C/T]GTGTGCACTGCTAGTTTGTTGAGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRYBG3 PubMed Links

Gene Details

Gene
CRYBG3
Gene Name
crystallin beta-gamma domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153605.3 Intron NP_705833.3
XM_005247117.4 Intron XP_005247174.1

View Full Product Details