Product Details

SNP ID
rs79547129
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:97993220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATATCTTGAGCCTAAACAAACACT[C/T]TCACTGGATTTTTCTGTCTCTTTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GABRR3 PubMed Links

Gene Details

Gene
GABRR3
Gene Name
gamma-aminobutyric acid type A receptor rho3 subunit (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105580.2 Intron NP_001099050.1

View Full Product Details