Product Details
- SNP ID
-
rs79977940
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:151294734 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGGATGATGTTTGGCAAAGACAAA[A/C]CAGCCATGATCACCCAAACACAAAC
- Phenotype
-
MIM: 606379
MIM: 611318
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GPR87
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2242353] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GPR87
- Gene Name
- G protein-coupled receptor 87
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_023915.3 |
873 |
Missense Mutation |
GGT,GTT |
G171V |
NP_076404.3 |
- Gene
- MED12L
- Gene Name
- mediator complex subunit 12 like
View Full Product Details