Product Details
- SNP ID
-
rs78536844
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:158670372 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAAAAATAGAATGAGAAAAGCAGA[C/T]GTTAAGAAGATGATTGATTGGGGAA
- Phenotype
-
MIM: 606639
MIM: 609305
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GFM1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs144626831] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GFM1
- Gene Name
- G elongation factor mitochondrial 1
- Gene
- LXN
- Gene Name
- latexin
View Full Product Details