Product Details

SNP ID
rs79811397
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:169840930 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGATGAAATAAAGACTTCTGTAA[A/G]AGGAGGCAGGAGAGATAACCACGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC31 PubMed Links

Gene Details

Gene
LRRC31
Gene Name
leucine rich repeat containing 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277127.1 Intron NP_001264056.1
NM_001277128.1 Intron NP_001264057.1
NM_024727.3 Intron NP_079003.2
XM_011513158.2 Intron XP_011511460.1
XM_011513159.2 Intron XP_011511461.1
XM_011513160.2 Intron XP_011511462.1
XM_017007204.1 Intron XP_016862693.1
Gene
LRRIQ4
Gene Name
leucine rich repeats and IQ motif containing 4
There are no transcripts associated with this gene.

View Full Product Details