Product Details
- SNP ID
-
rs77585458
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:170420671 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGGCCTCTGGCACACAATAGCTGC[A/G]GGCAGTGCTTGCCTGGTTTGCCTTA
- Phenotype
-
MIM: 601326
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CLDN11
PubMed Links
Gene Details
- Gene
- CLDN11
- Gene Name
- claudin 11
- Gene
- MIR6828
- Gene Name
- microRNA 6828
There are no transcripts associated with this gene.
View Full Product Details