Product Details

SNP ID
rs79691858
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:102398520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAATAATCTCCTAACTGGCCTTTC[C/T]CCTTTATATACTACCTCCAGAGACA
Phenotype
MIM: 615915
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ZPLD1 PubMed Links

Gene Details

Gene
ZPLD1
Gene Name
zona pellucida like domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175056.1 Intron NP_778226.1
XM_017005703.1 Intron XP_016861192.1
XM_017005704.1 Intron XP_016861193.1

View Full Product Details