Product Details

SNP ID
rs77148492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:183123806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTACATATATTATGTTAATTCA[A/T]CATATCTCAAATGTTGACTTTGAGT
Phenotype
MIM: 605883
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LAMP3 PubMed Links

Gene Details

Gene
LAMP3
Gene Name
lysosomal associated membrane protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014398.3 1750 UTR 3 NP_055213.2
XM_005247360.4 1750 Intron XP_005247417.1
XM_006713586.3 1750 Intron XP_006713649.1
XM_011512688.1 1750 Intron XP_011510990.1

View Full Product Details