Product Details

SNP ID
rs76019285
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184563888 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTATGTGTCACCTGTGTGTGTC[A/G]GGCCACAAGGCCCTTGTTGTGGGTG
Phenotype
MIM: 601839
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF2B5-AS1 PubMed Links

Gene Details

Gene
EIF2B5-AS1
Gene Name
EIF2B5 antisense RNA 1
There are no transcripts associated with this gene.

Gene
EPHB3
Gene Name
EPH receptor B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004443.3 Intron NP_004434.2

View Full Product Details