Product Details

SNP ID
rs76874736
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184710464 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTCAAATAGCTAAGCAAATGAAG[A/G]TACATGCCAGTCTTTCTTCTTTTAT
Phenotype
MIM: 609267
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAGEF1 PubMed Links

Gene Details

Gene
MAGEF1
Gene Name
MAGE family member F1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022149.4 1585 UTR 3 NP_071432.2

View Full Product Details