Product Details

SNP ID
rs77545529
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:6801917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATACATGCCACCATTACCTTCCA[C/G]ATAAAGTTTGTATCCCTCACTGTGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KIAA0232 PubMed Links

Gene Details

Gene
KIAA0232
Gene Name
KIAA0232
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100590.1 Intron NP_001094060.1
NM_014743.2 Intron NP_055558.2
XM_005248041.4 Intron XP_005248098.1
XM_011513598.2 Intron XP_011511900.1
XM_017008860.1 Intron XP_016864349.1
XM_017008861.1 Intron XP_016864350.1
XM_017008862.1 Intron XP_016864351.1
XM_017008863.1 Intron XP_016864352.1

View Full Product Details